Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-5-(trifluoromethyl)pyridine, 98+%

Catalog No. AC241780250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC241780250 25 g Glass bottle
AC241780050 5 g Glass bottle
AC241781000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC241780250 Supplier Thermo Scientific Chemicals Supplier No. 241780250
Add to Cart

CAS: 52334-81-3 | C6H3ClF3N | 181.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52334-81-3
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
MDL Number MFCD00042225
InChI Key JFZJMSDDOOAOIV-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl pyridine, 2,5-ctf, pyridine, 2-chloro-5-trifluoromethyl, ccris 6430, 5-trifluoromethyl-2-chloropyridine, unii-c9g66d966b, 2-chloro-5-trifluoropyridine, 2-chloro-5 trifluoromethyl pyridine
PubChem CID 92342
IUPAC Name 2-chloro-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC=C(Cl)N=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 29°C to 33°C
Color White to Yellow
Boiling Point 152°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: reacts at high temperature with strong acids and a
Formula Weight 181.51
Percent Purity 98+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2-chloro-5-(trifluoromethyl)pyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 257-856-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.