Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-ethoxypyridine, 98%, Thermo Scientific™

Catalog No. AAB2498409
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2498409 10 g
AAB2498418 50 g
AAB2498430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2498409 Supplier Thermo Scientific Chemicals Supplier No. B2498409

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42144-78-5
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.597
MDL Number MFCD00023465
InChI Key AMSLPXHLKHZWBJ-UHFFFAOYSA-N
PubChem CID 98563
IUPAC Name 2-chloro-6-ethoxypyridine
SMILES CCOC1=NC(=CC=C1)Cl

Specifications

Density 1.17
Refractive Index 1.519
Quantity 10 g
Synonym pyridine, 2-chloro-6-ethoxy, acmc-209jns, 6-chloro-2-ethoxypyridine, 2-chloro-6-ethoxypyridine, 2-chloro-6-ethoxy-pyridine, pyridine,2-chloro-6-ethoxy, 2-chloranyl-6-ethoxy-pyridine
Formula Weight 157.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-ethoxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 255-681-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.