Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methoxypyridine, 98%

Catalog No. AAL0599714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0599714 25 g
AAL0599706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0599714 Supplier Thermo Scientific Chemicals Supplier No. L0599714
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 17228-64-7 | C6H6ClNO | 143.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17228-64-7
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD00006265
InChI Key VAVGOGHLNAJECD-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-6-methoxy, 2-methoxy-6-chloropyridine, 6-chloro-2-methoxypyridine, 2-chloro-6-methoxy-pyridine, 2-chlor-6-methoxypyridin, pubchem13194, acmc-1cffb, 6-methoxy-2-chloropyridine, 2-chloro-6-methoxy pyridine, 2-chloro-6-methyloxy pyridine
PubChem CID 87009
IUPAC Name 2-chloro-6-methoxypyridine
SMILES COC1=NC(=CC=C1)Cl

Specifications

Density 1.207
Boiling Point 185°C to 186°C
Flash Point 76°C (168°F)
Refractive Index 1.528
Quantity 25 g
Beilstein 1364500
Formula Weight 143.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methoxypyridine
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 241-264-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.