Learn More
Sigma Organic Chemistry 2-Chloro-7- methylthio | 1G | MFCD02049247
Supplier: Sigma Organic Chemistry CBR021421G

2-Chloro-7- methylthio , 1G
About this Item:
MDL #: MFCD02049247
Molecular Weight: 113.2
UNSPSC Code: 12352200
Chemical Formula: C7H15N
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.