Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroacetophenone, 98%, Thermo Scientific Chemicals

Catalog No. AAA1074922
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1074922 100 g
AAA1074936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1074922 Supplier Thermo Scientific Chemicals Supplier No. A1074922
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-27-4
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000933
InChI Key IMACFCSSMIZSPP-UHFFFAOYSA-N
Synonym 2-chloroacetophenone, phenacyl chloride, omega-chloroacetophenone, chloroacetophenone, tear gas, mace lacrimator, alpha-chloroacetophenone, ethanone, 2-chloro-1-phenyl, mace, 1-chloroacetophenone
PubChem CID 10757
IUPAC Name 2-chloro-1-phenylethan-1-one
SMILES ClCC(=O)C1=CC=CC=C1

Specifications

Melting Point 52°C to 56°C
Density 1.324
Boiling Point 243°C to 244°C
Flash Point 118°C (244°F)
Odor Irritating
Quantity 100 g
UN Number UN1697
Beilstein 507950
Merck Index 14,2115
Formula Weight 154.6
Percent Purity 98%
Chemical Name or Material 2-Chloroacetophenone
Show More Show Less
Hazard Category H301-H310-H314
Hazard Statement GHS H Statement
H300-H331-H319-H317-H335
Fatal if swallowed.
Toxic if inhaled.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P260-P262-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROACETOPHENONE, SOLID
EINECSNumber 208-531-1
RTECSNumber AM6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.