Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroadenosine hemihydrate, 97%, Thermo Scientific™

Catalog No. AC230290100
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC230290100 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC230290100 Supplier Thermo Scientific Chemicals Supplier No. 230290100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81012-94-4
Molecular Formula C10H14ClN5O5
Molecular Weight (g/mol) 319.702
MDL Number MFCD00149351
InChI Key CSVJEVDOHIWPDO-GWTDSMLYSA-N
Synonym 2-chloroadenosine hydrate, 2-chloroadenosine hemidydrate
PubChem CID 70699968
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-2-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol;hydrate
SMILES C1=NC2=C(N1C3C(C(C(O3)CO)O)O)N=C(N=C2N)Cl.O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 10 mg
Formula Weight 310.69
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroadenosine hemihydrate, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.