Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzohydrazide 98.0+%, TCI America™
SDP

Catalog No. C211625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C211625G 25 g
C21165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C211625G Supplier TCI America Supplier No. C211625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5814-05-1
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.60
MDL Number MFCD00007597
InChI Key KPPNLSKVTKSSTG-UHFFFAOYSA-N
Synonym 2-chlorobenzhydrazide, 2-chlorobenzoic hydrazide, 2-chlorobenzoic acid hydrazide, 2-chlorobenzene-1-carbohydrazide, benzoylhydrazine, o-chloro, benzoic acid, 2-chloro-, hydrazide, 2-chlorobenzenecarbohydrazide, chlorobenzohydrazide, o-chlorobenzhydrazide, 2clphcon2
PubChem CID 79883
IUPAC Name 2-chlorobenzohydrazide
SMILES C1=CC=C(C(=C1)C(=O)NN)Cl

Specifications

Melting Point 118°C
Quantity 25 g
Formula Weight 170.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorobenzohydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.