Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzoic acid, 98%

Catalog No. AC159240020
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159240020 2 kg Plastic bottle
AC159240050 5 g Glass Bottle
AC159241000 100 g Plastic bottle
AC159245000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC159240020 Supplier Thermo Scientific Chemicals Supplier No. 159240020
Add to Cart

CAS: 118-91-2 | C7H5ClO2 | 156.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-91-2
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00002412
InChI Key IKCLCGXPQILATA-UHFFFAOYSA-N
Synonym o-chlorobenzoic acid, benzoic acid, o-chloro, benzoic acid, 2-chloro, 2-cba, kyselina o-chlorbenzoova, chlorobenzoic acid, ocba, benzoic acid, chloro, 2-chloro-benzoic acid, chlorobenzoate
PubChem CID 8374
ChEBI CHEBI:30793
IUPAC Name 2-chlorobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 139.0°C to 143.0°C
Color White
Boiling Point 285.0°C
Flash Point 173°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula ClC6H4CO2H
Quantity 2 kg
Beilstein 09, 334
Merck Index 15, 2125
Solubility Information Solubility in water: soluble in hot water. Other solubilities: freely soluble in alcohol and ether
Formula Weight 156.57
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chlorobenzoic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clo
EINECSNumber 204-285-4
RTECSNumber DG4976000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.