Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethanesulfonyl Chloride 95.0+%, TCI America™
SDP

Catalog No. C1142100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1142100G 100 g
C114225G 25 g
C1142500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C1142100G Supplier TCI America Supplier No. C1142100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1622-32-8
Molecular Formula C2H4Cl2O2S
Molecular Weight (g/mol) 163.012
MDL Number MFCD00007461
InChI Key VHCSBTPOPKFYIU-UHFFFAOYSA-N
Synonym 2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride
PubChem CID 15385
IUPAC Name 2-chloroethanesulfonyl chloride
SMILES C(CCl)S(=O)(=O)Cl

Specifications

Color Yellow
Boiling Point 203°C
Quantity 100 g
UN Number 2927
Formula Weight 163.01
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloroethanesulfonyl Chloride
RTECSNumber KI8060000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.