Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl Chloroformate 98.0+%, TCI America™
SDP

Catalog No. C1052100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1052100G 100 g
C105225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1052100G Supplier TCI America Supplier No. C1052100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 627-11-2
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.96
MDL Number MFCD00000646
InChI Key SVDDJQGVOFZBNX-UHFFFAOYSA-N
Synonym 2-chloroethyl chloroformate, chloroethyl chloroformate, carbonochloridic acid, 2-chloroethyl ester, chloroformic acid 2-chloroethyl ester, formic acid, chloro-, 2-chloroethyl ester, chloroethylchloroformate, 2-chloroethylchloroformate, unii-4d32h07z9i, beta-chloroethyl chlorocarbonate, chloroformic acid, beta-chloroethyl ester
PubChem CID 12305
IUPAC Name 2-chloroethyl carbonochloridate
SMILES ClCCOC(Cl)=O

Specifications

Color Yellow
Boiling Point 154°C
Quantity 100 g
UN Number 2742
Formula Weight 142.96
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Chloroformate
EINECSNumber (2)-3629
RTECSNumber LQ5950000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.