Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl isocyanate, 98%

Catalog No. AC221190010
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221190010 1 mL Glass bottle
AC221190050 5 mL Glass bottle
AC221190250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221190010 Supplier Thermo Scientific Chemicals Supplier No. 221190010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1943-83-5 | C3H4ClNO | 105.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1943-83-5
Molecular Formula C3H4ClNO
Molecular Weight (g/mol) 105.52
MDL Number MFCD00002043
InChI Key BCMYXYHEMGPZJN-UHFFFAOYSA-N
Synonym 2-chloroethyl isocyanate, 2-chloroethylisocyanate, chloroethylisocyanate, ethane, 1-chloro-2-isocyanato, 2-chlorethylisokyanat, 2-chloro ethyl isocyanate, isocyanic acid, 2-chloroethyl ester, 1-chloro-2-isocyanato-ethane, 2-chlorethylisokyanat czech, .beta.-chloroethylisocyanate
PubChem CID 16035
IUPAC Name 1-chloro-2-isocyanatoethane
SMILES C(CCl)N=C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2370g/mL
Boiling Point 135°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4460 to 1.449
Linear Formula ClCH2CH2NCO
Quantity 1 mL
Beilstein 04, III, 245
Specific Gravity 1.237
Solubility Information Solubility in water: reacts
Formula Weight 105.52
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl isocyanate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-734-4
RTECSNumber NQ8450000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.