Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethylvinyl Ether Solution, 5000μg/mL in methanol, Ultra Scientific

Catalog No. USEPA1016
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1016 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1016 Supplier Agilent Technologies Supplier No. EPA10161
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 110-75-8 , 67-56-1
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.549
InChI Key DNJRKFKAFWSXSE-UHFFFAOYSA-N
PubChem CID 8074
IUPAC Name 1-chloro-2-ethenoxyethane
SMILES C=COCCCl

Specifications

CAS Max % 99.36, 0.63
Analyte or Component Names methanol,carbon disulfide
Element Methanol, Carbon Disulfide
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material 2-Chloroethylvinyl Ether Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.