Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorohexane 98.0+%, TCI America™
SDP

Catalog No. C089025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C089025G 25 g
C08905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C089025G Supplier TCI America Supplier No. C089025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 638-28-8
Molecular Formula C6H13Cl
Molecular Weight (g/mol) 120.62
MDL Number MFCD00039351
InChI Key GLCIPJOIEVLTPR-UHFFFAOYNA-N
Synonym 2-Hexyl Chloride
PubChem CID 12521
IUPAC Name 2-chlorohexane
SMILES CCCCC(C)Cl

Specifications

Color Colorless
Boiling Point 122°C
Quantity 25 g
UN Number 1993
Formula Weight 120.62
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorohexane
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.