Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloromethyl-1,3-dioxolane 95.0+%, TCI America™
SDP

Catalog No. C132125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C132125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C132125G Supplier TCI America Supplier No. C132125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2568-30-1
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD00043402
InChI Key IKZOMJGRWIOEDP-UHFFFAOYSA-N
PubChem CID 75727
IUPAC Name 2-(chloromethyl)-1,3-dioxolane
SMILES C1COC(O1)CCl

Specifications

Color Yellow
Boiling Point 158°C
Quantity 25 g
Formula Weight 122.55
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloromethyl-1,3-dioxolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.