Learn More
Sigma Organic Chemistry 2-(Chloromethyl)pyridine hydrochloride | 25G | 6959-47-3 | MFCD00012811
Supplier: Sigma Organic Chemistry 16270125G
CAS #: 6959-47-3
MDL #: MFCD00012811
Purity: >98 %
UNSPSC Code: 12352100
Molecular Weight: 164.03
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.