Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloropyrazine, 98%

Manufacturer:  Alfa Aesar™ A1010822

Quantity 100g

 View more versions of this product

Catalog No. AAA1010822


  Check Availability
Add to cart

Specifications

Description

Applications
2-Chloropyrazine is used in cardiotonic, in biological, drug, flavoring and perfumery industry.

Solubility
Soluble in Methanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Description & Specifications

Chemical Identifiers

14508-49-7
114.54
UN1993
2-Chloropyrazine
C4H3ClN2
MFCD00006124
107019

Specifications

57°C (134°F)
100g
C4H3ClN2
MFCD00006124
107019
98%
153° to 154°C
14508-49-7
114.54
UN1993
2-Chloropyrazine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit