Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloropyridine-3-boronic acid pinacol ester, 97%

Catalog No. AC430660010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430660010 1 g Glass bottle
AC430660050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC430660010 Supplier Thermo Scientific Chemicals Supplier No. 430660010
Add to Cart

CAS: 452972-11-1 | C11H15BClNO2 | 239.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452972-11-1
Molecular Formula C11H15BClNO2
Molecular Weight (g/mol) 239.51
MDL Number MFCD04039873
InChI Key XFZFMAUUZHBQSS-UHFFFAOYSA-N
Synonym 2-chloropyridine-3-boronic acid pinacol ester, 2-chloro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-chloro-3-pyridineboronic acid pinacol ester, 2-chloropyridin-3-ylboronic acid pinacol ester, 2-chloropyridine-3-boronicacidpinacolester, 2-chloropyridine-3-boronic acid, pinacol ester, 2-chloro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-chloro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem16529
PubChem CID 12067060
IUPAC Name 2-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CC=CN=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 34.0°C to 39.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 239.61
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloropyridine-3-boronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.