Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloropyridine, 99%

Catalog No. AC110010025
Change view
Click to view available options
Quantity:
100 mL
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110010025 2.5 L Glass bottle
AC110011000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC110010025 Supplier Thermo Scientific Chemicals Supplier No. 110010025

CAS: 109-09-1 | C5H4ClN | 113.544 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-09-1
MDL Number MFCD00006228
InChI Key OKDGRDCXVWSXDC-UHFFFAOYSA-N
Synonym o-chloropyridine, pyridine, 2-chloro, alpha-chloropyridine, .alpha.-chloropyridine, unii-8hmd45ayej, 2-chloro-pyridine, ccris 1724, 8hmd45ayej, 6-chloro-pyridine, dsstox_cid_4810
PubChem CID 7977
ChEBI CHEBI:39174
IUPAC Name 2-chloropyridine
SMILES C1=CC=NC(=C1)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -46.0°C
Color Colorless to Yellow
Density 1.2090g/mL
Boiling Point 168.0°C to 170.0°C
Flash Point 64°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H4ClN
Refractive Index 1.5310 to 1.5330
Linear Formula (C5H4N)Cl
Quantity 2.5 L
Beilstein 20, 230
Specific Gravity 1.209
Solubility Information Solubility in water: 27g/L (20°C). Other solubilities: >50g/100mL ethanol,soluble in ether and alcohol
Molecular Weight (g/mol) 113.55
Formula Weight 113.55
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chloropyridine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Very toxi
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 203-646-3
RTECSNumber US5950000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.