Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorothiophene, 98+%, Thermo Scientific™

Catalog No. AAA103900B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA103900B 1000 g
AAA1039018 50 g
AAA1039030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA103900B Supplier Thermo Scientific Chemicals Supplier No. A103900B

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the preparation of 5-phenylthiophene derivative and N-arylhydroxamic acids.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-43-5
Molecular Formula C4H3ClS
Molecular Weight (g/mol) 118.578
MDL Number MFCD00005421
InChI Key GSFNQBFZFXUTBN-UHFFFAOYSA-N
Synonym 2-thienyl chloride, thiophene, 2-chloro, 2-chloro thiophene, 2-chlorthiophen, chlorothiophene, thienyl chloride, 5-chlorothiophene, thiophene, chloro, sfpdadhhryszzp@, pubchem7376
PubChem CID 7299
IUPAC Name 2-chlorothiophene
SMILES C1=CSC(=C1)Cl

Specifications

Melting Point -72°C
Density 1.285
Boiling Point 127°C to 129°C
Flash Point 22°C (71°F)
Odor Unpleasant
Refractive Index 1.548
Quantity 1000 g
UN Number UN1992
Beilstein 104652
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 118.59
Percent Purity ≥98%
Chemical Name or Material 2-Chlorothiophene
Show More Show Less
Hazard Category H225-H301+H311+H331
Hazard Statement GHS H Statement
H225-H301-H311-H331
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 202-505-3
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.