Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 2% Cholesterol Diet, Pellet

Complete rat feed with 2% cholesterol added.

Supplier:  MP Biomedicals™ 0290469120

 View more versions of this product

Catalog No. MP290469120


Only null left
Add to Cart

Description

Description

  • 2% Cholesterol
  • For rodent diet
  • Pellets
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Animal Diet
Rodent diet
20 kg
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.