Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyano-4'-methylbiphenyl, 98+%

Catalog No. AAB2127314
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2127314 25 g
AAB2127306 5 g
AAB2127322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2127314 Supplier Thermo Scientific Chemicals Supplier No. B2127314
Add to Cart

CAS: 114772-53-1 | C14H11N | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114772-53-1
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00151805
InChI Key ZGQVZLSNEBEHFN-UHFFFAOYSA-N
Synonym 2-cyano-4'-methylbiphenyl, 4'-methyl-2-cyanobiphenyl, 4'-methylbiphenyl-2-carbonitrile, 2'-cyano-4-methylbiphenyl, 2-p-tolyl benzonitrile, 2-4-methylphenyl benzonitrile, 2-cyano-4'-methyl biphenyl, 4'-methyl-1,1'-biphenyl-2-carbonitrile, unii-o417ti15ta, 4'-methyl-2-cyano diphenyl
PubChem CID 145512
IUPAC Name 2-(4-methylphenyl)benzonitrile
SMILES CC1=CC=C(C=C1)C1=CC=CC=C1C#N

Specifications

Melting Point 49°C to 53°C
Density 1.17
Boiling Point >320°C
Quantity 25 g
UN Number UN3077
Beilstein 3605954
Formula Weight 193.26
Percent Purity ≥98%
Chemical Name or Material 2-Cyano-4'-methylbiphenyl
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H372-H361-H302
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 422-310-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.