Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyanoethyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™
SDP

Catalog No. C197425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C197425G 25 g
C19745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C197425G Supplier TCI America Supplier No. C197425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 106-71-8
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.13
MDL Number MFCD00013823
InChI Key AEPWOCLBLLCOGZ-UHFFFAOYSA-N
Synonym Acrylic Acid 2-Cyanoethyl Ester, 3-Acryloyloxypropionitrile
PubChem CID 7825
IUPAC Name 2-cyanoethyl prop-2-enoate
SMILES C=CC(=O)OCCC#N

Specifications

Color Yellow
Boiling Point 108°C
Quantity 25 g
UN Number 3276
Formula Weight 125.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyanoethyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-3416
RTECSNumber AT1500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.