Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Cyanoethyl N,N-diisopropylchlorophosphoramidite

Catalog No. 302309250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-230-9250MG 250 mg
30-230-95G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-230-9250MG Supplier Sigma Aldrich Supplier No. 302309250MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-cyanoethyl n,n-diisopropylchlorophosphoramidite was employed:
  • For selective monophosphorylation of carbohydrates and nucleosides
  • For conversion of protected ribonucleosides to phosphoramidites
  • As phosphitylating reagent for 3α-hydroxyl groups in the synthesis of oligodeoxyribonucleotides
  • In a scalable, solution-phase oligonucleotide synthesis employing phosphoramidite chemistry and dmt-, ibu- and bz-protected monomers. intermediates were isolated by extraction without further purification

Specifications

CAS 89992-70-1
Density 1.061 g/mL (at 25°C (literature))
Molecular Formula C9H18ClN2OP
Linear Formula [(CH3)2 CH]2 NP(Cl)OCH2CH2CN
MDL Number MFCD00011544
Quantity 250 mg
Synonym Chloro(diisopropylamino)-beta-cyanoethoxyphosphine
Molecular Weight (g/mol) 236.68
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.