Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyanophenylacetonitrile 98.0+%, TCI America™
SDP

Catalog No. C224725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C224725G 25 g
C22475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C224725G Supplier TCI America Supplier No. C224725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3759-28-2
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00001895
InChI Key GKHSEDFDYXZGCG-UHFFFAOYSA-N
Synonym 2-cyanomethyl benzonitrile, homophthalonitrile, 2-cyanobenzyl cyanide, 2-cyanophenylacetonitrile, alpha-cyano-o-tolunitrile, o-cyanobenzyl cyanide, benzeneacetonitrile, 2-cyano, 2-cyanobenzeneacetonitrile, o-cyanophenyl acetonitrile, o-tolunitrile, alpha-cyano
PubChem CID 77368
IUPAC Name 2-(cyanomethyl)benzonitrile
SMILES N#CCC1=CC=CC=C1C#N

Specifications

Melting Point 80°C
Quantity 25 g
Formula Weight 142.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanophenylacetonitrile
RTECSNumber XV0930000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.