Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyanopyrimidine 98.0+%, TCI America™
SDP

Catalog No. C151825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C151825G 25 g
C15181G 1 g
C15185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C151825G Supplier TCI America Supplier No. C151825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14080-23-0
Molecular Formula C5H3N3
Molecular Weight (g/mol) 105.10
MDL Number MFCD00160513
InChI Key IIHQNAXFIODVDU-UHFFFAOYSA-N
Synonym 2-cyanopyrimidine, 2-pyrimidinecarbonitrile, 2-cyano-pyrimidine, 2-cyano pyrimidine, cyanopyrimidine, provastatin, pyrimidinecarbonitrile, zlchem 283, pubchem7042, pyrimidin-2-carbonitrile
PubChem CID 2757979
IUPAC Name pyrimidine-2-carbonitrile
SMILES N#CC1=NC=CC=N1

Specifications

Melting Point 41°C
Quantity 25 g
Formula Weight 105.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanopyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.