Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyclohexylethanol, 99%

Catalog No. AAB207820B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB207820B 1000 g
AAB2078218 50 g
AAB2078230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB207820B Supplier Thermo Scientific Chemicals Supplier No. B207820B
Add to Cart

CAS: 4442-79-9 | C8H16O | 128.215 g/mol

Used as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agent.

Solubility
Soluble in alcohol and dipropylene glycol. Soluble in water (1750 mg/L at 25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4442-79-9
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00001525
InChI Key QJQZRLXDLORINA-UHFFFAOYSA-N
Synonym cyclohexaneethanol, cyclohexylethanol, cyclohexylethyl alcohol, 2-cyclohexylethan-1-ol, ethanol, 2-cyclohexyl, hexahydrophenylethyl alcohol, unii-653aok9puo, 2-hydroxyethyl cyclohexane, .beta.-cyclohexylethyl alcohol, 2-cyclohexyl-ethanol
PubChem CID 20508
IUPAC Name 2-cyclohexylethanol
SMILES C1CCC(CC1)CCO

Specifications

Melting Point -20°C
Density 0.92
Boiling Point 206°C to 207°C
Flash Point 86°C (186°F)
Refractive Index 1.466
Quantity 1000 g
Beilstein 1848152
Solubility Information Soluble in alcohol and dipropylene glycol. Soluble in water (1750mg/L at 25°C).
Formula Weight 128.22
Percent Purity 99%
Chemical Name or Material 2-Cyclohexylethanol
Show More Show Less
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312-H227
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
EINECSNumber 224-672-1
RTECSNumber KK3528000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.