Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyclopenten-1-one 96.0+%, TCI America™
SDP

Catalog No. C079425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C079425ML 25 mL
C07945ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C079425ML Supplier TCI America Supplier No. C079425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 930-30-3
Molecular Formula C5H6O
Molecular Weight (g/mol) 82.10
MDL Number MFCD00001401
InChI Key BZKFMUIJRXWWQK-UHFFFAOYSA-N
Synonym 2-cyclopenten-1-one, 2-cyclopentenone, cyclopentenone, cyclopent-2-enone, cyclopenten-3-one, 2-cyclopentene-1-one, cyclopent-2-ene-1-one, unii-q0u2igf9ck, 3-cyclopenten-2-one, q0u2igf9ck
PubChem CID 13588
IUPAC Name cyclopent-2-en-1-one
SMILES O=C1CCC=C1

Specifications

Color Yellow
Boiling Point 154°C
Quantity 25 mL
UN Number 1224
Formula Weight 82.10
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclopenten-1-one
EINECSNumber (3)-4493
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.