Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyclopentenone, 98%

Catalog No. AC111570250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111570250 25 g Glass bottle
AC111570050 5 g Glass bottle
AC111571000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111570250 Supplier Thermo Scientific Chemicals Supplier No. 111570250
Add to Cart

CAS: 930-30-3 | C5H6O | 82.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-30-3
Molecular Formula C5H6O
Molecular Weight (g/mol) 82.10
MDL Number MFCD00001401
InChI Key BZKFMUIJRXWWQK-UHFFFAOYSA-N
Synonym 2-cyclopenten-1-one, 2-cyclopentenone, cyclopentenone, cyclopent-2-enone, cyclopenten-3-one, 2-cyclopentene-1-one, cyclopent-2-ene-1-one, unii-q0u2igf9ck, 3-cyclopenten-2-one, q0u2igf9ck
PubChem CID 13588
IUPAC Name cyclopent-2-en-1-one
SMILES O=C1CCC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 0.9800g/mL
Boiling Point 150°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4800 to 1.482
Linear Formula C5H6(=O)
Quantity 25 g
Beilstein 07, I, 45
Specific Gravity 0.98
Solubility Information Solubility in water: almost insoluble. Other solubilities: very soluble in ethanol,ether
Formula Weight 82.1
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2-Cyclopentenone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-213-
Recommended Storage May darken on storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.