Learn More
Sonoco ThermoSafe Brands 2 Day Certis™ Platform
Certis™ series offers robust universal temperature control for temperature sensitive goods. The Certis solutions are material neutral, incorporating leading edge technologies to provide the lowest total cost of ownership
Supplier:  Sonoco ThermoSafe Brands C13F2U
 View more versions of this product
Catalog No. 03-600-275
Only null left
Add to Cart
Description
Description
  • 2 Day solutions for a variety of temperature ranges
  • All shippers are prequalified systems saving assembly time and space and optimize dim weight and freight cost.
  • Use the same common components across temperature ranges and durations
  • Same pack-out configurations across temperature ranges
  • Mistake-proof packout
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 EACH
19.5 x 18.5 x 17.5
11 x 9.5 x 7.5
13L
Shipper, gel packs, pads, payload box, and packout instructions
2 days; 48 hours
Frozen Temperature
Expanded Polystyrene Foam
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.