Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Decanol, 98%

Catalog No. AC331600250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC331600250 25 g
AC331600050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC331600250 Supplier Thermo Scientific Chemicals Supplier No. 331600250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1120-06-5 | C10H22O | 158.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-06-5
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.28
MDL Number MFCD00004594
InChI Key ACUZDYFTRHEKOS-UHFFFAOYSA-N
Synonym 2-decanol, 2-hydroxydecane, 2-decyl alcohol, methyl-n-octylcarbinol, decanol-2, methyl-n-octyl carbinol, acmc-1bovc, chembl46477
PubChem CID 14254
IUPAC Name decan-2-ol
SMILES CCCCCCCCC(C)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -5°C
Color Colorless
Density 0.8200g/mL
Boiling Point 211°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4330 to 1.435
Linear Formula CH3(CH2)7CH(OH)CH3
Quantity 25 g
Beilstein 01, I, 213
Specific Gravity 0.82
Solubility Information Solubility in water: insoluble in water
Formula Weight 158.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Decanol, 95%
Show More Show Less
EINECSNumber 214-296-6
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.