Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Deoxy-5-(hydroxymethyl)cytidine 98.0+%, TCI America™
SDP

Catalog No. D4220200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4220200MG 200 mg
D422050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4220200MG Supplier TCI America Supplier No. D4220200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7226-77-9
Molecular Formula C10H15N3O5
Molecular Weight (g/mol) 257.246
MDL Number MFCD09842113
InChI Key HMUOMFLFUUHUPE-CSMHCCOUSA-N
Synonym 5-hmdC
PubChem CID 98007919
IUPAC Name 4-amino-1-[(2S,4R,5S)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(hydroxymethyl)pyrimidin-2-one
SMILES C1C(C(OC1N2C=C(C(=NC2=O)N)CO)CO)O

Specifications

Melting Point 205°C
Quantity 200 mg
Formula Weight 257.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxy-5-(hydroxymethyl)cytidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.