Learn More
Trilink Bio Technologies Inc 2'-Deoxy-P-nucleoside-5'-Triphosphate, 1 µmole
SDP
Supplier:  Trilink Bio Technologies Inc N20371
Encompass
Molecular Formula C11H18N3O14P3, Purity Specification: ≥90% by AX-HPLC, Molecular Weight: 509.2 g/mol, Salt Form: Lithium
Catalog No. NC0606404
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.