Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Deoxyadenosine-5'-diphosphate Trisodium Salt, ≥97%, MP Biomedicals™

Catalog No. ICN15079425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15079425 25 mg
ICN15079480 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15079425 Supplier MP Biomedicals, Inc Supplier No. 0215079425
Add to Cart
Add to Cart

Chemical Identifiers

CAS 72003-83-9
Molecular Formula C10H15N5Na2O9P2
Molecular Weight (g/mol) 457.18
MDL Number MFCD00083610,MFCD00083610
InChI Key PMCITFSCGIPAKY-UHFFFAOYNA-N
Synonym 2'-deoxyadenosine-5'-diphosphate disodium salt
PubChem CID 131858105
IUPAC Name [({[5-(6-amino-9H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid disodium
SMILES [Na].[Na].NC1=C2N=CN(C3CC(O)C(COP(O)(=O)OP(O)(O)=O)O3)C2=NC=N1

Specifications

Color White
Assay Percent Range ≥97%
Quantity 25 mg
Formula Weight 397.2
Physical Form Amorphous Powder
Chemical Name or Material 2'-Deoxyadenosine-5'-diphosphate Trisodium Salt
EINECSNumber 276-280-5
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.