Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Deoxyadenosine-5'-monophosphate, 98%

Catalog No. AAJ6440206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6440206 5 g
AAJ6440203 1 g
AAJ6440214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6440206 Supplier Thermo Scientific Chemicals Supplier No. J6440206
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 653-63-4 | C10H14N5O6P | 331.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in 1N ammonium hhydroxide (50 mg/ml) and water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-63-4
Molecular Formula C10H14N5O6P
Molecular Weight (g/mol) 331.23
MDL Number MFCD00005753
InChI Key KHWCHTKSEGGWEX-RRKCRQDMSA-N
Synonym damp, 2'-deoxyadenosine-5'-monophosphate, 2'-deoxyadenosine 5'-monophosphate, 2'-deoxyadenylic acid, deoxyadenylic acid, deoxy-amp, 2'-deoxy-amp, 2'-deoxyadenosine 5'-phosphate, 2'-damp, deoxyadenosine monophosphate
PubChem CID 12599
ChEBI CHEBI:17713
IUPAC Name [(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](COP(O)(O)=O)O3)C2=NC=N1

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in 1N ammonium hhydroxide (50mg/ml) and water.
Formula Weight 331.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2'-Deoxyadenosine-5'-monophosphate
EINECSNumber 211-503-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.