Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 2'-Deoxythymidine 5'-Monophosphate Disodium Salt, >98% Purity, 1 g

Catalog No. 502704590
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4590 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4590 Supplier Abcam Supplier No. AB1465321G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deoxyribonucleotide substrate for molecular biology applications with >98% purity.

  • Purity: >98%
  • Molecular Weight: 366.2 g/mol
  • Applications: Deoxyribonucleotide substrate for molecular biology applications
  • Composition: 100% Thymidine-5'-monophosphoric acid, disodium

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Deoxyribonucleotide
Form Powder
Shipping Condition Ambient
Substrate Deoxyribonucleotide
Content And Storage Store at -20°C.
Buffer 100% thymidine-5'-monophosphoric acid, disodium
Formulation 100% THYMIDINE-5'-MONOPHOSPHORIC ACID, Disodium
Quantity 1 g
For Use With (Application) Molecular biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.