Learn More
Abcam 2'-Deoxythymidine 5'-Monophosphate Disodium Salt, >98% Purity, 250 mg
Description
- Molecular weight: 366.2 g/mol
- Purity: >98%
- Composition: 100% Thymidine-5¹-monophosphoric acid, disodium
- Application: Substrate in molecular biology applications
- Storage temperature: -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | 2'-Deoxythymidine 5'-monophosphate |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Buffer | 100% thymidine-5'-monophosphoric acid, disodium |
| Content And Storage | Store at -20°C. |
| Formulation | 100% THYMIDINE-5'-MONOPHOSPHORIC ACID, Disodium |
| Quantity | 250 mg |
| For Use With (Application) | Molecular biology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.