Learn More
Abcam 2'-Deoxyuridine-5'-Monophosphate Disodium Salt, >98% Purity, Solid, 250 mg
Description
- Molecular weight: 352.1 g/mol
- Purity: >98%
- Enzyme substrate: Reference substrate for thymidylate synthetase (TS) (EC 2.1.1.45)
- Application: Used for the de novo production of dTMP and in studies of dUMP analogs in potential chemotherapy applications
- Form: Solid
- CAS Number: 42155-08-8
- Storage buffer: Constituents: 100% 2'-Deoxy-5'-uridylic acid disodium salt
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Thymidylate Synthetase |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Substrate | dUMP |
| Buffer | 100% 2'-Deoxy-5'-uridylic acid disodium salt |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% 2'-Deoxy-5'-uridylic acid disodium salt |
| Quantity | 250 mg |
| For Use With (Application) | Thymidylate synthetase enzymatic studies, studies of dUMP analogs in chemotherapy applications |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.