Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 2'-Deoxyuridine-5'-monophosphate disodium salt, 99%

Catalog No. AAJ6462703
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6462703 1 g
AAJ64627MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6462703 Supplier Thermo Scientific Chemicals Supplier No. J6462703
Add to Cart

CAS: 42155-08-8 | C9H13N2Na2O8P | 354.162 g/mol

2?-Deoxyuridine 5?-monophosphate (dUMP) is used by the enzyme thymidylate synthetase (TS) (EC 2.1.1.45) for the de novo production of dTMP. dUMP is used as the reference substrate in studies of dUMP analogues in potential chemotherapy applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyuridine 5′-monophosphate (dUMP) is used by the enzyme thymidylate synthetase (TS) (EC 2.1.1.45) for the de novo production of dTMP. dUMP is used as the reference substrate in studies of dUMP analogues in potential chemotherapy applications.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42155-08-8
Molecular Formula C9H13N2Na2O8P
Molecular Weight (g/mol) 354.162
MDL Number MFCD00065282
InChI Key CWTUMDATELIIAI-CDNBRZBRSA-N
Synonym 2'-deoxyuridine 5'-monophosphate disodium salt
PubChem CID 129893468
IUPAC Name [(2R,3S,5R)-5-(2,4-dioxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
SMILES C1C(C(OC1N2C=CC(=O)NC2=O)COP(=O)(O)O)O.[Na].[Na]

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 352.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Deoxyuridine-5'-monophosphate disodium salt
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 255-687-1
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.