Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 2'-Deoxyuridine-5'-monophosphate Disodium Salt, Solid, >98% Purity, 1 g

Catalog No. 502704827
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4827 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4827 Supplier Abcam Supplier No. AB1462241G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reference substrate for studies of dUMP analogs in potential chemotherapy applications.

  • Molecular Weight: 352.1 g/mol
  • Purity: >98%
  • Applications: Reference substrate used by thymidylate synthetase (EC 2.1.1.45) for the de novo production of dTMP; used in studies of dUMP analogs for potential chemotherapy applications
  • Form: Solid
  • Storage Buffer: 100% 2'-Deoxy-5'-uridylic acid disodium salt

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Thymidylate Synthetase
Buffer 100% 2'-Deoxy-5'-uridylic acid disodium salt
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Substrate dUMP
Formulation 100% 2'-Deoxy-5'-uridylic acid disodium salt
Quantity 1 g
For Use With (Application) Thymidylate synthetase enzyme studies, dUMP analog research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.