Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. D50391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50391G 1 g
D50395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50391G Supplier TCI America Supplier No. D50391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 564483-19-8
Molecular Formula C29H45P
Molecular Weight (g/mol) 424.653
MDL Number MFCD06411306
InChI Key SACNIGZYDTUHKB-UHFFFAOYSA-N
Synonym 2-di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl, di-tert-butyl 2',4',6'-triisopropyl-1,1'-biphenyl-2-yl phosphine, tbuxphos, tert-butyl xphos, t-butylxphos, 2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, t-bu xphos, di-t-bu-xphos, di-tert-butyl 2',4',6'-triisopropylbiphenyl-2-yl phosphine, 2-di-t-butylphosphino-2',4',6'-triisopropylbiphenyl
PubChem CID 11618717
IUPAC Name ditert-butyl-[2-[2,4,6-tri(propan-2-yl)phenyl]phenyl]phosphane
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)C2=CC=CC=C2P(C(C)(C)C)C(C)(C)C)C(C)C

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 1 g
Formula Weight 424.65
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Di-tert-butylphosphino-2′,4′,6′-triisopropylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.