Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dicyclohexylphosphino)biphenyl 98.0+%, TCI America™
SDP

Catalog No. D33881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33881G 1 g
D33885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33881G Supplier TCI America Supplier No. D33881G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 247940-06-3
Molecular Formula C24H31P
Molecular Weight (g/mol) 350.486
MDL Number MFCD01862441
InChI Key LCSNDSFWVKMJCT-UHFFFAOYSA-N
Synonym 2-dicyclohexylphosphino biphenyl, cyjohnphos, 1,1'-biphenyl-2-yldicyclohexylphosphine, 2-biphenyl dicyclohexylphosphine, 2-biphenylyl dicyclohexylphosphine, biphenyl-2-yldicyclohexylphosphine, dicyclohexyl 2-phenylphenyl phosphane, cyclohexyl johnphos, dicyclohexyl-2-phenylphenyl phosphane, 2-dicyclohexylphosphino-biphenyl
PubChem CID 2734216
IUPAC Name dicyclohexyl-(2-phenylphenyl)phosphane
SMILES C1CCC(CC1)P(C2CCCCC2)C3=CC=CC=C3C4=CC=CC=C4

Specifications

Melting Point 104°C
Color White
Quantity 1 g
Formula Weight 350.49
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Dicyclohexylphosphino)biphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.