Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Diethylaminoethylchloride hydrochloride, 99.5%, Thermo Scientific Chemicals

Catalog No. AC114030025
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114030025 2.5 kg Plastic drum
AC114030050 5 g Glass Bottle
AC114032500 250 g Glass Bottle
AC114030010 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC114030025 Supplier Thermo Scientific Chemicals Supplier No. 114030025
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869-24-9
Molecular Formula C6H15Cl2N
Molecular Weight (g/mol) 172.09
MDL Number MFCD00012519
InChI Key RAGSWDIQBBZLLL-UHFFFAOYSA-N
Synonym 2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
PubChem CID 13363
IUPAC Name 2-chloro-N,N-diethylethanamine;hydrochloride
SMILES [H+].[Cl-].CCN(CC)CCCl

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point 208.0°C to 212.0°C
Color White to Beige
Loss on Drying 1% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99.5%
Packaging Plastic drum
Linear Formula (C2H5)2NCH2CH2Cl·HCl
Quantity 2.5 kg
Beilstein 04, III, 240
Solubility Information Solubility in water: 2000g/L (20°C)
Formula Weight 172.1
Percent Purity 99.5%
Physical Form Crystalline Powder
Chemical Name or Material 2-Diethylaminoethylchloride hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 212-786-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.