Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Difluoromethoxy)benzylamine, TCI America™
SDP

Catalog No. D48381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D48381G Supplier TCI America Supplier No. D48381G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 243863-36-7
Molecular Formula C8H9F2NO
Molecular Weight (g/mol) 173.163
MDL Number MFCD00236229
InChI Key GGXXUJUQXAOYPK-UHFFFAOYSA-N
Synonym 2-difluoromethoxy benzylamine, 2-difluoromethoxy phenyl methanamine, 1-2-difluoromethoxy phenyl methanamine, benzenemethanamine, 2-difluoromethoxy, 2-difluoromethoxy phenyl methylamine, acmc-20a5tk, 2-difluoromethoxy-benzylamine, 2-difluoromethoxy benzyl amine
PubChem CID 1514062
IUPAC Name [2-(difluoromethoxy)phenyl]methanamine
SMILES C1=CC=C(C(=C1)CN)OC(F)F

Specifications

Color Yellow
Quantity 1 g
UN Number 2735
Formula Weight 173.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Difluoromethoxy)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.