Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Difluoromethyl)benzimidazole 98.0+%, TCI America™
SDP

Catalog No. D47631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47631G 1 g
D47635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47631G Supplier TCI America Supplier No. D47631G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 705-09-9
Molecular Formula C8H6F2N2
Molecular Weight (g/mol) 168.147
MDL Number MFCD00463015
InChI Key PURNIHSRWGYONZ-UHFFFAOYSA-N
PubChem CID 597703
IUPAC Name 2-(difluoromethyl)-1H-benzimidazole
SMILES C1=CC=C2C(=C1)NC(=N2)C(F)F

Specifications

Melting Point 157°C
Quantity 1 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Difluoromethyl)benzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.