Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Diisopropylaminoethyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AAB226620E
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB226620E 2500 g
AAB2266222 100 g
AAB2266236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB226620E Supplier Thermo Scientific Chemicals Supplier No. B226620E
Add to Cart

2-Diisopropylaminoethyl chloride hydrochloride is used as an intermediate in pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Diisopropylaminoethyl chloride hydrochloride is used as an intermediate in pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

Solubility
Soluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4261-68-1
Molecular Formula C8H19Cl2N
Molecular Weight (g/mol) 200.15
MDL Number MFCD00012496
InChI Key IUSXYVRFJVAVOB-UHFFFAOYSA-N
Synonym 2-diisopropylaminoethyl chloride hydrochloride, 2-diisopropylamino ethyl chloride hydrochloride, unii-6rmr38fceq, 2-chloroethyldiisopropylamine hydrochloride, 6rmr38fceq, 2-propanamine, n-2-chloroethyl-n-1-methylethyl-, hydrochloride, 2-chloroethyldiisopropylammonium chloride, 2-chloroethyl diisopropylamine hydrochloride, n-chloroethyl diisopropylamine hydrochloride, beta-chloroethyl diisopropylamine hydrochloride
PubChem CID 77942
SMILES [H+].[Cl-].CC(C)N(CCCl)C(C)C

Specifications

Melting Point 131°C to 134°C
Quantity 2500 g
UN Number UN2811
Beilstein 3551603
Sensitivity Hygroscopic
Solubility Information Soluble in water.
IUPAC Name hydrogen (2-chloroethyl)bis(propan-2-yl)amine chloride
Formula Weight 200.15
Percent Purity 98%
Chemical Name or Material 2-Diisopropylaminoethyl chloride hydrochloride
Hazard Category H301+H311-H315-H319-H330-H335
Hazard Statement GHS H Statement
H300-H310-H330-H318-H315-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 224-238-1
RTECSNumber TX1521900
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.