Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dimethylamino)-6-propionylnaphthalene 98.0+%, TCI America™
SDP

Catalog No. D46781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46781G 1 g
D4678200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46781G Supplier TCI America Supplier No. D46781G

Chemical Identifiers

CAS 70504-01-7
Molecular Formula C15H17NO
Molecular Weight (g/mol) 227.31
MDL Number MFCD00056615
InChI Key MPPQGYCZBNURDG-UHFFFAOYSA-N
Synonym Prodan
PubChem CID 107729
ChEBI CHEBI:51909
IUPAC Name 1-[6-(dimethylamino)naphthalen-2-yl]propan-1-one
SMILES CCC(=O)C1=CC=C2C=C(C=CC2=C1)N(C)C

Specifications

Melting Point 138°C
Color Yellow
Quantity 1 g
Formula Weight 227.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Dimethylamino)-6-propionylnaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.