Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dimethylamino)vinyl 3-Pyridyl Ketone 98.0+%, TCI America™
SDP

Catalog No. D39491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39491G 1 g
D39495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39491G Supplier TCI America Supplier No. D39491G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 55314-16-4
Molecular Formula C10H12N2O
Molecular Weight (g/mol) 176.219
MDL Number MFCD00115180
InChI Key MZLRFUCMBQWLNV-FNORWQNLSA-N
Synonym 1-3-pyridyl-3-dimethylamino-2-propen-1-one, e-3-dimethylamino-1-pyridin-3-yl prop-2-en-1-one, 3-dimethylamino-1-pyridin-3-yl prop-2-en-1-one, 2e-3-dimethylamino-1-pyridin-3-yl prop-2-en-1-one, e-3-dimethylamino-1-pyridin-3-yl-propenone, dimethylaminopyridinylpropenone, 3-dimethylamino-1-3-pyridyl-2-propen-1-one, 3-dimethylamino-1-3-pyridinyl-2-propene-1-one, 3-3-dimethylamino acryloyl pyridine, e-3-dimethylamino-1-pyridin-3-ylprop-2-en-1-one
PubChem CID 5369165
IUPAC Name (E)-3-(dimethylamino)-1-pyridin-3-ylprop-2-en-1-one
SMILES CN(C)C=CC(=O)C1=CN=CC=C1

Specifications

Melting Point 71°C
Quantity 1 g
Formula Weight 176.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Dimethylamino)vinyl 3-Pyridyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.