Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Dimethylaminoethanethiol hydrochloride, 95%, Thermo Scientific Chemicals

Catalog No. AC115811000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC115811000 100 g
AC115810050 5 g
AC115810250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC115811000 Supplier Thermo Scientific Chemicals Supplier No. 115811000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13242-44-9
Molecular Formula C4H11NS·HCl
Molecular Weight (g/mol) 141.67
MDL Number MFCD00012633
InChI Key NRVFDGZJTPCULU-UHFFFAOYSA-N
Synonym 2-dimethylamino ethanethiol hydrochloride, captamine hydrochloride, meda, captamine hcl, n,n-dimethylcysteamine hydrochloride, dimethylaminoethanethiol hydrochloride, 2-dimethylaminoethanethiol hydrochloride, n-dimethylcysteamine hydrochloride, 2-mercaptoethyl dimethylammonium chloride, unii-m43ax41u87
PubChem CID 25798
IUPAC Name 2-(dimethylamino)ethanethiol;hydrochloride
SMILES CN(C)CCS.Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 154°C to 159°C
Assay Percent Range 94% min. (Iodimetry)
Packaging Glass Bottle
Linear Formula (CH3)2NCH2CH2SH·HCl
Quantity 100 g
Solubility Information Solubility in water: soluble in water
Formula Weight 141.67
Percent Purity 95%
Chemical Name or Material 2-Dimethylaminoethanethiol hydrochloride, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 236-221-6
RTECSNumber KJ1051000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.