Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals

Catalog No. AC115820050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115820050 5 g Glass Bottle
AC115821000 100 g Plastic bottle
AC115825000 500 g Plastic bottle
AC115820025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC115820050 Supplier Thermo Scientific Chemicals Supplier No. 115820050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4584-46-7 , 107-99-3
Molecular Formula C4H10ClN·HCl
Molecular Weight (g/mol) 144.05
MDL Number MFCD00012516
InChI Key LQLJZSJKRYTKTP-UHFFFAOYSA-N
Synonym 2-dimethylaminoethyl chloride hydrochloride, 2-chloro-n,n-dimethylethanamine hydrochloride, 2-dimethylamino ethyl chloride hydrochloride, 2-chloroethyl dimethylamine hydrochloride, 2-chloro-n,n-dimethylethylamine hydrochloride, dimethylaminoethyl chloride hydrochloride, 2-chloroethyldimethylamine hydrochloride, 1-chloro-2-dimethylaminoethane hydrochloride, 2-dimethylamino ethylchloride hydrochloride, n-2-chloroethyl dimethylamine hydrochloride
PubChem CID 11636816
ChEBI CHEBI:78153
IUPAC Name 2-chloro-N,N-dimethylethanamine;hydrochloride
SMILES CN(C)CCCl.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 203.0°C to 210.0°C
Color Brown-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula (CH3)2NCH2CH2Cl·HCl
Quantity 5 g
Beilstein 04, 133
Solubility Information Solubility in water: 2000g/L (20°C). Other solubilities: soluble in alcohol (20°C)
Formula Weight 144.05
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Dimethylaminoethyl chloride hydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a
EINECSNumber 224-970-1
RTECSNumber KQ9020000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.