Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Dodecylhexadecan-1-ol 93.0+%, TCI America™
SDP

Catalog No. D5322250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5322250G 250 g
D532225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5322250G Supplier TCI America Supplier No. D5322250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 72388-18-2
Molecular Formula C28H58O
Molecular Weight (g/mol) 410.77
MDL Number MFCD27665210
InChI Key DEMBLPGWNXUBIQ-UHFFFAOYNA-N
Synonym 2-Dodecylhexadecyl Alcohol
PubChem CID 3018340
IUPAC Name 2-dodecylhexadecan-1-ol
SMILES CCCCCCCCCCCCCCC(CO)CCCCCCCCCCCC

Specifications

Melting Point 35°C
Color White
Quantity 250 g
Formula Weight 410.77
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Dodecylhexadecan-1-ol
EINECSNumber (2)-0217
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.